Learn More
Ansell RINGERS™ R080 Waterproof Impact Gloves
Catalog No. 17004297 Shop All Ansell Products
Change view
Click to view available options
Size:
8
9
10
11
12
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-004-297 12
17-004-293 8
17-004-294 9
17-004-295 10
17-004-296 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-004-297 Supplier Ansell Supplier No. 08012
Add to Cart
Add to Cart
This item is not returnable. View return policy
Waterproof cut-resistant impact gloves, with a nitrile-dipped palm for excellent dexterity.
  • The nitrile dipped palm and waterproof barrier combines protection against impact hazards with 360 cut resistance and liquid protection for a wide range of applications
  • A breathable knit shell developed with Ringers' F3 Technology™
  • Patented TPR design provide superior dexterity and comfort
  • The waterproof liner helps keep hands dry while maintaining excellent dexterity
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Nitrile
Color Gray
Cut Resistance Rating ANSI A6
Hand Specific
Latex Free Yes
Material HPPE
Product Type Cut-resistant Gloves
Size 12
Style Ringers™ R080
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.