Learn More
Ansell RINGERS™ R167 TPR-designed Impact Gloves
Catalog No. 17004298 Shop All Ansell Products
Change view
Click to view available options
Size:
8
9
10
11
12
13
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-004-298 8
17-004-299 9
17-004-300 10
17-004-301 11
17-004-302 12
17-004-303 13
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-004-298 Supplier Ansell Supplier No. 16708
Add to Cart
Add to Cart
This item is not returnable. View return policy
Touchscreen-compatible impact and cut protection gloves, for hand safety and ease of use.
  • Certified ANSI/ISEA 138 Level 1 Flexible knuckle TPR design with two detached fingers Durable synthetic
  • Developed with Ringers' F3 Technology™ and patented TPR design for superior dexterity and comfort
  • Touchscreen compatible index, middle and thumb tips lessen the need to remove gloves
  • High visibility and hook and loop pull tab closure provides added security on the job
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Leather
Color Yellow
Cut Resistance Rating ANSI A3
Latex Free Yes
Size 8
Style Ringers™ R-167
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.