Learn More
Description
- Optimized Construction: Up to 30% lighter, and thinner than comparable gloves.
- Lightweight Impact protection: Level 1 TPR shields hands without restricting movement. Enhanced comfort, dexterity, and reduced hand fatigue.
- Extreme level cut-resistance: ANSI A9 and EN ISO F+ ratings deliver reliable safety without compromising comfort or dexterity.
- Increased Grip & Durability: Microfoam nitrile coating reduces replacements costs and delivers reliable grip for extended wear.
- Reinforced for Longevity: Thumb Crotch Reinforcement (TCR) strengthens high-wear zones to extend glove life and cut replacement costs.
- Touchscreen Compatible: Allows the wearer to work multiple touchscreen devices while keeping gloves on.
Specifications
Specifications
| Certifications/Compliance | ANSI/ISEA 105-2024 Abrasion Level 5, ANSI/ISEA 105-2024 Impact 138-1, FDA Approved |
| Certifications/Compliance (Europe) | EN ISO 21420:2020, Approved Contact with Food, Dermatest GmbH |
| Coated Area | Palm |
| Construction | Knit |
| Coating Material | Nitrile |
| Color | Black |
| Cut Resistance Rating | ANSI A9 |
| Hand | Specific |
| Latex Free | Yes |
| Material | HPPE, Spandex, Lycra, Nitrile |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.