Learn More
Ansell™ RINGERS™ R668
Catalog No. 17720636 Shop All Ansell Products
Change view
Click to view available options
Size:
9
10
11
12
13
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-720-636 9
17-720-637 10
17-720-638 11
17-720-639 12
17-720-640 13
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-720-636 Supplier Ansell™ Supplier No. 66809
Add to Cart
Add to Cart
Premium leather with 360° cut, level 2 impact and G-100 Thinsulate™ for cold weather.
  • Thermal goatskin leather: G-100 Thinsulate lined for warmth and durability
  • Superior cut-resistant lining: For impact gloves with 360° cut resistance
  • Patented TPR design: Knuckles, thumb and finger-length impact protection
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Cut A6, ANSI ABR 4, ANSI/ISEA 105-2006 PUN 3
Coated Area Uncoated
Construction Cut and Sewn
Description Medium-duty impact gloves are fully lined with high-quality goatskin leather.
Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A6
Hand Specific
Latex Free Yes
Material HPPE
Product Type Mechanical Protection Gloves
Size 9
Gender Unisex
Glove Cuff Color White
Glove Cuff Style Open
Grip Design Goatskin leather
Liner Material HPPE, Thinsulate, Polyester, Goatskin leather
Lining Lined
Model R668
Puncture Rating ANSI/ISEA 105-16 Level 3
Thumb Style Wing