Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ringers Rope Rescue Gloves

Catalog No. 19822752 Shop All Ansell Products
Encompass
Change view
Click to view available options
Color:
Black
Red
Size:
Small
Medium
Large
X-Large
2X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-822-752 Black Small
19-822-753 Black Medium
19-822-754 Black Large
19-822-757 Red Small
19-822-756 Black 2X-Large
19-822-755 Black X-Large
19-822-758 Red Medium
19-822-759 Red Large
19-822-760 Red X-Large
19-822-761 Red 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 19-822-752 Supplier Ringers Supplier No. 35308
Add to Cart
Add to Cart

Suited for all rope handling applications

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Neoprene, Synthetic Leather
Product Type Extrication Gloves
Size Small
For Use With (Application) Rescue, Rappelling, and SWAT Situations
Glove Cuff Style Hook and Loop
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.