Learn More
MBL International Corporation RIP-Assay Kit for microRNA
SDP
Supplier:  MBL International Corporation RN1005
Encompass_Preferred
Size:10 Assays; Target:RNP Complex; RIP-Assay Kit for microRNA is optimized to immunochemically isolate endogenous miRNAs, mRNAs and RBPs within mRNP complexes. The kit is designed to isolate cellular miRNAs that being incorporated into the RISC and/or to isolate unique group of miRNAs that bind to specific group of mRNAs encoding functionally related proteins.RIP-Assay Kit.
Catalog No. 50-510-83
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.