Learn More
Invitrogen™ RIP140 Polyclonal Antibody

Description
PA1-841 detects recombinant human Receptor-Interacting Protein 140 (RIP 140). PA1-841 has been successfully used in Western blot and immunoprecipitation procedures. By Western blot, this antibody detects two proteins at ~140 kDa representing human RIP 140 expressed in rabbit reticulocyte lysate. The PA1-841 immunogen is a synthetic peptide corresponding to residues G(718) N P T K G R V K K K E K T P L R D(735) of human RIP140. This immunizing peptide (Cat. # PEP-030) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | RIP140 |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | NRIP1 |
| Gene Accession No. | P48552 |
| Gene Alias | ERAP 140; FLJ77253; NRIP1; Nuclear factor RIP140; nuclear receptor interacting protein 1; nuclear receptor-interacting protein 1; OTTHUMP00000083036; OTTHUMP00000083037; Receptor-interacting protein 140; RIP140 |
| Gene Symbols | NRIP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.