Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIP2/RICK Mouse, Unlabeled, Clone: 25, BD

Catalog No. BDB612348 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612348 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612348 Supplier BD Biosciences Supplier No. 612348
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Members of the TNFR family (TNFRs, DRs, Fas, lymphotoxin-β-receptor, CD40, CD30, and OX-40) regulate a variety of cellular responses, such as cell activation, proliferation, differentiation, NF-κB activation, and apoptosis. Signaling through TNFR family members involves several families of receptor-associated proteins. RIP and RIP2 (RICK/Cardiak) are ser/thr kinase adaptor molecules that associate with TNFR complexes. Both RIPs contain homologous N-teriminal ser/thr kinase domains, but RIP contains a C-terminal death domain, while RIP2 contains a C-terminal caspase activation and recruitment domain (CARD) similar to those found in IAPs. Both RIP and RIP2 can activate NF-κB and cause cell death. RIP2 is recruited to TNFRs through interactions with TRAF1, TRAF5, and TRAF6, and RIP2 activation of NF-κB requires IKKα, IKKβ, and IKKγ. In addition, RIP2 can be activated through interactions with Ras-activated Raf1, and RIP2 can activate ERK1 and ERK2. Thus, RIP proteins may regulate TNFR signaling through both ser/thr kinase activity and interaction with the apoptotic machinery.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen RIP2/RICK
Applications Western Blot
Classification Monoclonal
Clone 25
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human RIP2/RICK aa. 333-532
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.