Learn More
Boston Bioproducts Inc RIPA Buffer (with EDTA) - 250ml
Supplier: Boston Bioproducts Inc BP115D250ML
The RIPA Buffer is used for rapid, efficient cell lysis and solubilization of proteins from adherent and suspension cultured mammalian cells and tissue samples. With the combined action of its ionic detergents and EDTA, the RIPA buffer is extremely useful for disruption of all proteins within cellular compartments, including nuclear, membrane, and mitochondria. This buffer is compatible with many downstream applications, such as SDS-PAGE, immunoprecipitation, Western blot, and ELISA.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.