Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIPPLY3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563139
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563139 Supplier Invitrogen™ Supplier No. PA563139
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVQATIDFYD DESTESASEA EEPEEGPPPL HLLPQEVGGR QENGPGGKGR DQGINQGQRS SGGGDHWGEG PLPQGVSSRG GKCSSSK Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 29%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RIPPLY3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RIPPLY3
Gene Accession No. P57055
Gene Alias Down syndrome critical region gene 6; down syndrome critical region protein 6; DSCR6; Protein ripply3; ripply transcriptional repressor 3; RIPPLY3; ripply3 homolog
Gene Symbols RIPPLY3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RIPPLY3. Recombinant protein control fragment (Product #RP-91797).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53820
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.