Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RITA
GREENER_CHOICE

Catalog No. 244310
Click to view available options
Quantity:
1 mg
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

MDM2-p53 interaction inhibitor

Anti-tumor agent that binds wild-type p53 (Kd = 1.5 nM) preventing p53-MDM2 (HDM2) interaction. Induces p53 accumulation and stimulates apoptosis in tumor cell lines expressing wild-type p53 in vitro and in vivo. Inhibits HPV-E6-mediated proteasomal degradation. Suppresses expression of cervical carcinoma HeLa xenografts in vivo.

Chemical Identifiers

CAS 213261-59-7
Molecular Formula C14H12O3S2

Specifications

Quantity 10 mg
Formula Weight 292.37
Purity >97%
Molecular Weight (g/mol) 292.367
Chemical Name or Material RITA
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.