Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Rite-On Frosted Thick Slide

Catalog No. 12518101 Shop All Epredia Products
Change view
Click to view available options
Corner Style:
90°
Frosted:
2 Sides, Double Sandblasted
White Epoxy Ink Frosted End, 1 Side
White Epoxy Ink Frosted End, 1 Side, with GOLD SEAL Logo Printed Key
Thickness (Metric):
1 mm
1.2 mm
Length (Metric):
75 mm
76 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Corner Style Frosted Thickness (Metric) Length (Metric)
12-518-101 90° White Epoxy Ink Frosted End, 1 Side 1 mm 75 mm
12-518-104 90° 2 Sides, Double Sandblasted 1.2 mm 76 mm
12-518-104C 90° White Epoxy Ink Frosted End, 1 Side, with GOLD SEAL Logo Printed Key 1 mm 75 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-518-101 Supplier Epredia™ Supplier No. 3050002

Label slides quickly and easily with Epredia™ Gold Seal™ Rite-On™ and Ultra Frost™ Frosted Microscope slides, made from hard, colorless, noncorrosive glass.

All finished slides are sealed with a desiccant in an air tight, lint-free, hermetically sealed Moistur-Barrier™ package, keeping slides dry during shipping and storage. Slides work well in hematology, histology, cytology, microbiology and urinalysis; as well as other laboratory and research applications.

  • Smooth marking area for effortless writing, automated printing, or printed labels
  • Each case features an inner barrier-bag sealed with desiccant packs to protect from moisture contamination
  • Slide boxes convert into convenient slide storage files
  • Reduced iron content for low self-fluorescence
  • Manufactured from pure, water-white glass
  • Precleaned for immediate use
  • Part No. 3032-002 is used for fluorescent antibody examination

Specifications

Length (Metric) 75 mm
Edges Ground
Width (Metric) 25 mm
Thickness (Metric) 1 mm
Frosted White Epoxy Ink Frosted End, 1 Side
Precleaned Yes
Type Microscope Slide
Material Glass
Color White
Corner Style 90°
Description Rite-On Frosted Slide
Length (English) 3 in.
Quantity 72 Slides/Box, 50 Boxes/Case
Width (English) 1 in.
Dimensions (L x W) 3 x 1 in. (75 x 25 mm)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.