Description
- Water resistant 4-in-1 reversible windbreaker polyester jacket
- Weatherproof elastic band at wrists
- Removable zip-out sleeves — converts Class 3 jacket to Class 2 vest
- Reverses from hi-vis lime to gray/black
- Silver reflective tape on hi-vis side
- Hi-vis reflective piping on gray/black side
- Bottom hem adjustment — bungee cord/toggle pull
- Mic tab on hi-vis side
Specifications
Specifications
| Size | 5X-Large |
| Material | Polyester |
| Color | Hi-vis Lime |
| Closure Type | Zipper |
| Certifications/Compliance | Meets ANSI/ISEA 107-2015; Type R - Class 3 with sleeves hi-vis side only; Type R - Class 2 without sleeves hi-vis side only |
| Coating | TPU Coating and Water Resistant Treatment |
| No. of Pockets | 3 per side |
| Sleeve Style | Removable, Zip-out |
| Tape Type | China Brand Silver Reflective Stripes |