Learn More
TARGETMOL CHEMICALS INC RK33 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T69705MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RK33 (RK 33) is a first-in-class small molecule inhibitor of DDX3 (a RNA helicase) and causes G1 cell cycle arrest induces apoptosis and promotes radiation sensitization in DDX3-overexpressing cells. purity: 97%
Catalog No. 50-257-6401