Learn More
RKI Cylinders for Multi-Gas Detectors
Catalog No. 19134474
Click to view available options
Material:
Aluminum
Catalog No. Material
19-134-474 Aluminum
1 options
Catalog No. 19-134-474 Supplier RKI Supplier No. 810154RK02
Aluminum cylinders for gas calibration
  • H2S (25 ppm), CO (50 ppm), CH4 (50%) and O2 (12%) in N2
  • 58L (15 gal.)
Specifications
Type Aluminum Cylinder
Includes 25ppm Hydrogen Sulphide/50ppm Carbon Monoxide/50% LEL Methane/12% 02 in Nitrogen
Material Aluminum