Learn More
Tocris Bioscience™ RLLFT-NH2
Catalog No. 33931
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
33-931 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 33-931 Supplier Tocris Bioscience™ Supplier No. 3393/1
This item is not returnable. View return policy
Control peptide for TFLLR-NH2 (Cat. No. 1464)
RLLFT-NH2 is a reversed amino acid sequence control peptide for TFLLR-NH2, a PAR1 selective agonist that significantly increases nociceptive threshold.
Active Analog also available.
Specifications
Quantity 1 mg