Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RMDN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552680
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552680 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552680 Supplier Invitrogen™ Supplier No. PA552680
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DEVSCETVKM GRKDSLDLEE EAASGASSAL EAGGSSGLED VLPLLQQADE LHRGDEQGKR EGFQLLLNNK LVYGSRQDFL WRLARAYSDM CELTEEVSEK KSYALDGKEE AEAALEKGDE SADCHLWYAV LCG Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 84%.

RMDN3 is a protein coding gene involved in cellular calcium homeostasis regulation. May participate in differentiation and apoptosis of keratinocytes. Overexpression induces apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RMDN3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RMDN3
Gene Accession No. Q96TC7
Gene Alias 1200015F23Rik; AI131757; Cerebral protein 10; FAM82A2; Fam82c; family with sequence similarity 82, member A2; family with sequence similarity 82, member C; FLJ10579; hRMD-3; hucep-10; microtubule-associated protein; Protein FAM82A2; Protein FAM82C; Protein tyrosine phosphatase-interacting protein 51; Ptpip51; regulator of microtubule dynamics 3; regulator of microtubule dynamics protein 3; RGD1308697; Rmd3; RMD-3; Rmdn3; TCPTP-interacting protein 51; tyrosine phosphatase interacting protein 51; UNQ3122/PRO10274
Gene Symbols RMDN3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RMDN3. Recombinant protein control fragment (Product #RP-90272).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55177
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.