Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RMI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595632 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595632 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595632 Supplier Invitrogen™ Supplier No. PA595632
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human T-47D whole cell, human HepG2 whole cell, human K562 whole cell, rat thymus tissue, mouse HEPA1-6 whole cell. IHC: human intestinal cancer tissue, human intestinal cancer tissue, rat liver tissue, human mammary cancer tissue, human mammary cancer tissue, mouse liver tissue. Flow: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

RMI2 is a protein coding gene, essential component of the RMI complex, a complex that plays an important role in the processing of homologous recombination intermediates. It is required to regulate sister chromatid segregation and to limit DNA crossover. Essential for the stability, localization, and function of BLM, TOP3A, and complexes containing BLM. In the RMI complex, it is required to target BLM to chromatin and stress-induced nuclear foci and mitotic phosphorylation of BLM.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RMI2
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene RMI2
Gene Accession No. Q3UPE3, Q96E14
Gene Alias A630055G03Rik; BLAP18; BLM-associated protein of 18 kDa; C16orf75; Gm537; Gm929; hRMI2; RecQ mediated genome instability 2; recQ-mediated genome instability protein 2; RGD1563163; RMI2; RMI2, RecQ mediated genome instability 2, homolog; RMI2, RecQ mediated genome instability 2, homolog (S. cerevisiae); similar to hypothetical protein MGC24665
Gene Symbols RMI2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence of human RMI2 (KMTDLSDNPIHESMWELEVEDLHRNIP).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116028, 223970, 497856
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.