Learn More
Medchemexpress LLC HY-19976 5mg Medchemexpress, RN-1747 CAS:1024448-59-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY199765MG
Encompass_Preferred
Medchemexpress, HY-19976 5mg RN-1747 CAS:1024448-59-6 RN-1747 is a selective transient receptor potential cation channel subfamily V member 4 (TRPV4) agonist, with EC50 values are 0.77 μM, 4.0 μM and 4.1 μM for hTRPV4, mTRPV4 and rTRPV4 respectively. RN-1747 also antagonizes TRPM8, with an IC50 of 4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0901
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.