Learn More
Active Motif RNA ChIP-IT Control Kit – Human
SDP
Supplier:  Active Motif 53025
Encompass_Preferred
Active Motif's RNA ChIP-IT Control Kit - Human provides human-specific reagents (positive control Suz12 rabbit polyclonal antibody, negative control rabbit IgG and lincSFPQ PCR primer mix) to verify the integrity of chromatin samples prepared for use with the RNA ChIP-IT Kit, which is designed to characterize the role of RNA in genome regulation. The control kit also make it easier to validate if your antibodies will function in RNA-ChIP.
Catalog No. NC1851552
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.