Learn More
Apexbio Technology LLC Lambda Exonuclease(Synonyms: Lambda Exo, Lambda exonuclease enzyme, λ exonuclease, λ exonuclease enzyme), 10000U.
Supplier: Apexbio Technology LLC K109710000
Lambda Exonuclease is a DNA-specific exonuclease enzyme that progressively degrades double-stranded DNA in a 5 3 direction preferentially initiating digestion at 5 -phosphorylated termini It displays minor activity toward single-stranded DNA and double-stranded DNA with non-phosphorylated ends but cannot start cleavage at internal nicks or gaps Lambda Exonuclease is commonly employed in molecular biology laboratories for the generation of single-stranded DNA templates DNA single-strand conformation polymorphism (SSCP) analysis selective degradation of linear plasmid forms while preserving supercoiled structures and rolling-circle amplification protocols
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.