Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences RNA-Conserve™ Solution
SCI_ED

Catalog No. S02034
Change view
Click to view available options
Quantity:
10 mL
100 mL
50 x 1.5 mL
5 x 10 mL
5 x 100 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02034 10 mL
S02033 5 x 10 mL
S02030 50 x 1.5 mL
S02032 100 mL
S02031 5 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S02034 Supplier G-Biosciences Supplier No. 7861687
Add to Cart
Add to Cart

RNA-Conserve™ Solution for preserving RNA.

  • RNA-Conserve™ is designed for the preservation of RNA in biological samples, including but not limited to; cells, mammalian tissues, bacteria, RNA-virus, yeast, plant tissues, biological fluids, field and test site collected specimens, swab kits, and so forth.
  • Purified RNA when stored in RNA-Conserve™ is protected from degradation and will remain intact during prolonged storage.
  • This formulation is a solution of non-toxic salts and preserving agents, optimized to preserve the integrity of purified RNA.
  • Inactivation of RNases eliminates the urgency for immediate processing of the biological samples or cryopreservation.
  • Samples may be stored or transported without degradation of RNA samples.

Specifications

Buffer Proprietary RNA Storage Buffer, Binding Buffer, Rebinding Buffer, Lysis Buffer, Tissue Extraction Buffer
Control Type RNA Control, Endogenous Control, ERCC RNA Control, Positive Control, Endogenous Control
Isolation Technology Enzymatic, Cell Lysis (Nonionic Detergent), Syringe Filter (Glass Fiber), Hybridization Capture, One-phase Extraction
Species All, E. coli, Human, Mouse, Rat
Sample Type Biological Samples
Product Type RNA Preservation Solution
Quantity 10 mL
Sterility Non-sterile
Target Viral RNA, Total RNA, miRNA, mRNA, Pathogen DNA/RNA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.