Learn More
RNA Polymerase II, Mouse anti-Human, Yeast, Clone: 8WG16, Millipore Sigma™
Shop All MilliporeSigma ProductsDescription
Anti-RNA Polymerase II, CTD, clone 8WG16, Cat. No. 05-952-I, is a mouse monoclonal antibody that detects DNA-directed RNA polymerase II subunit RPB1 and has been tested for use in Dot Blot, Chromatin Immunoprecipitation, Immunoprecipitation, Inhibition assay, and Western Blotting.
Specifications
Specifications
| Antigen | RNA Polymerase II |
| Applications | ChIP Assay, Dot Blot, Immunoprecipitation, Inhibition Assays, Western Blot |
| Classification | Monoclonal |
| Clone | 8WG16 |
| Conjugate | Unconjugated |
| Dilution | Western Blotting Analysis: A representative lot detected RNA Polymerase II, CTD in Western Blotting applications (Patturajan, M., et. al. (1998). J Biol Chem. 273(8):4689-94; Zhang, J., et. al. (1991). J Biol Chem. 266(4):2290-6).Dot Blot Analysis: A representative lot detected RNA Polymerase II, CTD in Dot Blot applications (Thompson, N.E., et. al. (1989). J Biol Chem. 264(19):11511-20).Immunoprecipitation Analysis: A representative immunoprecipitated RNA Polymerase II, CTD in Immunoprecipitation applications (Bhaumik, S.R., et. al. (2001). Genes Dev. 15(15):1935-45).Inhibition Analysis: A representative lot inhibited the interaction of RNA Polymerase II, CTD with the transcription complex. (Thompson, N.E., et. al. (1989). J Biol Chem. 264(19):11511-20). |
| Formulation | Purified mouse monoclonal antibody IgG2a in PBS without preservatives. |
| Gene Alias | DNA-directed RNA polymerase II subunit RPB1;EC: 2.7.7.6;RNA polymerase II subunit B1;DNA-directed RNA polymerase II subunit A;DNA-directed RNA polymerase III largest subunit;RNA-directed RNA polymerase II subunit RPB1 |
| Gene Symbols | POLR2A;POLR2 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.