Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Promega™ RNA Wash Solution (RWA)

Catalog No. PRZ3091 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
58.8 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRZ3091 58.8 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRZ3091 Supplier Promega Supplier No. Z3091
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RNA Wash Solution (RWA) is a component of the SV and SV 96 Total RNA Isolation System, PureYield RNA Midiprep System and ReliaPrep miRNA Cell and Tissue Miniprep System for preparation of RNA from tissues and cultured cells.

  • Reagent Used In RNA Purification Protocols
  • Component of Promega RNA purification kits available for purchase separately

Specifications

Content And Storage 15°C to 30°C
Description RNA Wash Solution (RWA)
Quantity 58.8 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.