Learn More
Agilent Technologies RNACE-IT RIBONUCLEASECCKTAIL
Supplier:  Agilent Technologies 400720
Encompass
RNace-It Ribonuclease Cocktail 6000 U Contains: RNase A (2 mg/mL) nonspecific RNase from Aspergillus oryzae (4 units/microliter) Applications Any application where it is necessary to degrade RNA molecules
Catalog No. 50-125-651
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.