Learn More
Boston Bioproducts Inc Phosphate Buffered Saline, PBS (10X, RNase Free) - 500ml
Supplier:  Boston Bioproducts Inc BM220DR500ML
Encompass_Preferred
Phosphate Buffered Saline (PBS) is a versatile buffer solution widely used due to its isotonic and non-toxic nature to cells. With a pH range of 7.0 to 7.4, PBS mimics physiological osmolarity and ion concentrations, making it ideal for applications including protein purification, Western blotting and immunoassays, sample dilution, and as a transport medium. Packaged as a 10X concentration and tested for RNase, this solution can be diluted to support its application.
Catalog No. NC9853968
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.