Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences RNase Inhibitor (Murine)
SCI_ED

Catalog No. S03139
Change view
Click to view available options
Quantity:
20,000 Units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03139 20,000 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03139 Supplier G-Biosciences Supplier No. 7861978
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RNase Inhibitor (Murine) is used for first strand cDNA synthesis, RT-qPCR.

  • RNase Inhibitor is a recombinant protein of murine origin which specifically inhibits RNases A, B and C.
  • Moreover, it does not inhibit polymerase activity when mix with Taq DNA Polymerase and Reverse Transcriptase.

Specifications

Enzyme RNase Inhibitor
Sensitive to Heat Inactivation Yes
Quantity 20,000 Units
Product Type RNase Inhibitor
Research Category PCR, RNA Research, DNA Repair, Drug Development, Toxicology and Drug Metabolism
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.