Learn More
RNase R, 400 U, G-Biosciences

Description
- It is a 3-5' exoribonuclease that efficiently digests linear RNA molecules, leaving lariat, circular and double-stranded RNAs with short (<7 nt) 3' overhangs intact.
Specifications
Specifications
| Packaging | Vial |
| Enzyme | RNase |
| Sensitive to Heat Inactivation | Yes |
| Product Type | RNase R |
| Research Category | RNA Research, Genome Editing |
| Product Line | Thermo Scientific |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.