Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ RNaseP Verification Plate, 384-well

Catalog No. 4432500
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 384-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-325-00 1 x 384-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-325-00 Supplier Applied Biosystems™ Supplier No. 4432500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 384-Well RNaseP Verification Plate is used to verify your real-time PCR instrument with 384-Well Block

The 384-Well RNaseP Verification Plate is used to verify your real-time PCR instrument with 384-Well Block. Confirm the performance of your instrument by running the RNaseP assay. Please see your instrument user guide for full instructions on use.

The 384-Well RNaseP Verification Plate is preloaded with the necessary reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each well contains preloaded reaction mix (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye-labeled probe) and template.

The verification plate must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, it is recommended that the performance of your instrument be verified every six months.

TRUSTED_SUSTAINABILITY

Specifications

Format 384-well Rnase P Verification Plate
Content And Storage Store at –20°C ± 5°C.

Calibrated Dye Proprietary Mix
For Use With (Equipment) ViiA 7 Dx, QuantStudio Dx, QuantStudio 5 Dx, QuantStudio 7 Pro Dx
Product Type Instrument Verification Plate
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 1 x 384-well Plate
Calibration Type Instrument Verification Plate
PCR Method qPCR

For In Vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.