Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva RNAspin 96 RNA Isolation Kit

Catalog No. 45001757 Shop All Cytiva Products
Change view
Click to view available options
4 x 96 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
45-001-757 4 x 96 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-757 Supplier Cytiva Supplier No. 25050075
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Protocol and sufficient quantities of the following to carry out the number of preps indicated: DNase 1, RNase-free water, binding plates, 1.5mL tubes, wash plates, square well blocks, vacuum elution plates and buffers

For fast and efficient purification in a 96-well format

  • 96 samples parallel-processed via either centrifugation or vacuum with high reproducibility in under 70 minutes
  • Can be used to isolate total RNA from cultured cells, tissue, bacteria or yeast
  • DNase I treatment removes genomic DNA resulting in high-quality total RNA product
  • Isolated RNA is of sufficient quality (A260/A280 = 1.8-2.2) and quantity (up to 100μg) for multiple downstream experiments: quantitative reverse transcription polymerase chain reaction (QRT-PCR), primer extension and RNase protection assays
  • Integrated wash plate eliminates risk of contamination
  • Accessory tissue plate allows tissue samples to be processed
  • Sample size: up to 30mg for animal tissue, up to 2 x 106 cells for cell culture (vaccum); 1x 107 cells for cell culture (centrifuge); Elution volume: 50μ-130μL

TRUSTED_SUSTAINABILITY

Specifications

Description illustra RNAspin 96 Kit
Format 96-well Plate
Isolation Technology Organic Extraction
For Use With (Application) Used to reproducibly (well-to-well, plate-to-plate) isolate total RNA from cultured cells, tissue, bacteria or yeastCan also be used in a fully automated flow with vacuum on common laboratory workstationsSuitable for direct use in downstream application
Includes Protocol and sufficient quantities of the following to carry out the number of preps indicated, DNase 1, RNase-free Water, binding plates, 1.5mL tubes, wash plates, square well blocks, vacuum elution plates and Buffers
No. of Reactions 4 x 96 Preps
Product Type illustra RNAspin 96 Kit
RNase 1.8-2.2
Sample Volume 10 mg tissue-30 mg tissue
Test Time 70 min/4 preps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.