Learn More
Invitrogen™ RNF133 Monoclonal Antibody (3D6)

Description
Peptide Sequence: FFHKNCIDPW ILPHGTCPIC KCDILKVLGI QVVVENGTEP LQVLMSNELP ETLSPSEEET NNEVSPAGTS DKVIHVEENP TSQNNDIQPH SVVEDVHPSP.
Specifications
Specifications
| Antigen | RNF133 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3D6 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Rnf133 |
| Gene Accession No. | Q8WVZ7 |
| Gene Alias | E3 ubiquitin-protein ligase RNF133; Goliath-related E3 ubiquitin-protein ligase 2; Greul2; RING finger protein 133; RING-type E3 ubiquitin transferase RNF133; Rnf133 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.