Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF138 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110304
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110304 Supplier Invitrogen™ Supplier No. PA5110304
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVTRRERACP ERALDLENIM RRFSGSCRCC SKKIKFYRMR HHYKSCKKYQ DEYGVSSVIP NFKISQDSVR SSNRSETSAS DNTETYQEDT SSSGHPTFKC PLCQESNFTR QRLLDHCNSN HLFQIVPVTC PICVSLPWGD PSQITRNFVS HLNQRHQFDY GEFVNLQLDE ETQYQTAVEE SFQVNM.

The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-DNA and protein-protein interactions. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF138
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Rnf138
Gene Accession No. Q8WVD3
Gene Alias 2410015A17Rik; 2810480D20Rik; E3 ubiquitin-protein ligase RNF138; hNARF; HSD4; HSD-4; NARF; nemo-like kinase associated ring finger protein; Nemo-like kinase-associated RING finger protein; NLK-associated RING finger protein; ring finger protein 138; ring finger protein 138, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase RNF138; Rnf138; Rsd4; RSD-4; STRIN; testicular tissue protein Li 165; Trif; Trif-d
Gene Symbols Rnf138
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 60-245 of mouse RNF138 (Q9CQE0).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51444
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.