Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF146 Recombinant Rabbit Monoclonal Antibody (PSH0-29)
GREENER_CHOICE

Catalog No. FEMA550800 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550800 Supplier Invitrogen™ Supplier No. MA550800
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Positive control: SH-SY5Y cell lysate, NIH/3T3 cell lysate, human testis tissue, human thyroid cancer tissue, NIH/3T3 cell, SH-SY5Y cell. Predicted band size: 39 kDa Subcellular Location: Cytoplasm, Nucleus.

RNF146 protein function is currently unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF146
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone PSH0-29
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene RNF146
Gene Accession No. Q9CZW6, Q9NTX7
Gene Alias 2610509H23Rik; dactylidin; E3 ubiquitin-protein ligase RNF146; iduna; RING finger protein 146; RING finger protein 146; E3 ubiquitin-protein ligase RNF146; RING-type E3 ubiquitin transferase RNF146; Rnf146
Gene Symbols RNF146
Host Species Rabbit
Immunogen Recombinant protein within full length human RNF146.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68031, 81847
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.