Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566433
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566433 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566433 Supplier Invitrogen™ Supplier No. PA566433
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEKPEEDNDYH RSDEQVSICL ECNSSKLRGL KRKWIRCSAQ ATVLHLKKFI Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

Phosphorylase kinase is a hexadecameric enzyme that is comprised of four copies of four subunits that are encoded by four separate genes: PHKA, PHKB, PHKG, and PH kDa. This serine/threonine specific kinase converts glycogen phosphorylase b to glycogen phosphorylase a, resulting in the release of glucose-1-phophate from glycogen. PHKB (Phosphorylase b kinase regulatory subunit beta) is a 1093 amino acid subunit of phosphorylase kinase that, along with PHKA, has regulatory functions controlled by phosphorylation. Defects in the gene encoding PHKB are the cause of glycogen storage disease type 9B, which is also known as phosphorylase kinase deficiency of liver and muscle. This disease is characterized by a mild phenotype of hepatomegaly with only slightly elevated transaminase and plasma lipids, no clinical muscle involvement, and generally is correlated with a gradual improvement with increasing age. There are four isoforms of PHKB that are produced as a result of alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCGF3
Gene Accession No. Q3KNV8
Gene Alias 2310035N15Rik; AI662857; D630042K08Rik; D6S113E; DADB-17J1.3; DKFZp686D20235; DKFZp686N0336; DONG1; E430039C14; FLJ31942; FLJ36550; FLJ43813; FSH; FSRG1; KIAA9001; MGC129075 protein; MGC129615; MGC40413; NAT; PCGF3; polycomb group ring finger 3; polycomb group RING finger protein 3; ring finger protein 3; RING finger protein 3A; RING3; RNF3; RNF3A
Gene Symbols PCGF3
Host Species Rabbit
Immunogen Recombinant Human RNF3. Recombinant protein control fragment (Product #RP-107106).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10336
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.