Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561812
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561812 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561812 Supplier Invitrogen™ Supplier No. PA561812
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELALSLLQET PRNYELGDVV EAVRHSQDRA FLRRLLAQEC AVCGWALPHN RMQALTSCEC TICPDCFRQH FTIALKEKHI TDMVCPACGR PDLTDDTQLL SYFSTLDIQL RESLEPDAYA LFHKKLTEGV Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

Lyn proto oncogene (Lcl/Yes related novel protein tyrosine kinase) is encoded by the Lyn gene located on chromosome 8 in humans and belongs to the Src family kinase. It is a non-receptor tyrosine kinase that acts as a mediator in transducing cellular signaling. The most well studied expression of Lyn is in hematopoietic cell types, both lymphoid and myeloid cells with exclusion in T lymphocytes. It has also been observed that Lyn plays diverse roles such as in regulation of B cell receptor signaling, mast cell signaling and Epithelial - Mesenchymal Transition (EMT).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF31
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF31
Gene Accession No. Q96EP0
Gene Alias AL033293; BC031509; E3 ubiquitin-protein ligase RNF31; Flj10111; FLJ23501; HOIL-1-interacting protein; HOIP; mFLJ00217; MGC19975; Paul; putative Ariadne-like ubiquitin ligase; RING finger protein 31; RING-type E3 ubiquitin transferase RNF31; Rnf31; ZIBRA; Zinc in-between-RING-finger ubiquitin-associated domain protein
Gene Symbols RNF31
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF31. Recombinant protein control fragment (Product #RP-89026).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55072
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.