Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF32 purified MaxPab mouse Mouse anti-Human, Polyclonal, Abnova™

Catalog No. 89032207 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-207 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-207 Supplier Abnova Corporation Supplier No. H00140545B02P
Only null left
Add to Cart
Add to Cart

Mouse Polyclonal Antibody

Mouse polyclonal antibody raised against a full-length human RNF32 protein.

  • High specificity
  • High reactivity

Specifications

Antigen RNF32 purified MaxPab mouse
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human RNF32 protein.
Formulation PBS with no preservative; pH 7.4
Gene RNF32
Gene Accession No. BC028120.1
Gene Alias FKSG33/HSD15
Gene Symbols RNF32
Host Species Mouse
Immunogen RNF32 (AAH28120.1, 1 a.a. ∼ 256 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140545
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.