Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544865
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544865 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544865 Supplier Invitrogen™ Supplier No. PA544865
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AAAEEEDGGP EGPNRERGGA GATFECNICL ETAREAVVSV CGHLYCWPCL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.

The protein encoded by this gene contains a RING finger, which is a motif known to be involved in protein-protein interactions. This protein is a membrane-bound ubiquitin ligase. It can regulate cell motility by targeting paxillin ubiquitination and altering the distribution and localization of paxillin in cytoplasm and cell focal adhesions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RNF5
Gene Accession No. Q99942
Gene Alias 2410131O05Rik; AA407576; E3 ubiquitin-protein ligase RNF5; G16; HsRma1; ing finger protein 5 pseudogene; NG2; Protein G16; ram1 homolog; RING finger protein 5; ring finger protein 5, E3 ubiquitin protein ligase; RING5; RING-type E3 ubiquitin transferase RNF5; RMA1; RNF5
Gene Symbols RNF5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RNF5 (aa 2-51).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6048
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.