Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF90 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116170
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116170 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116170 Supplier Invitrogen™ Supplier No. PA5116170
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF90 Polyclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1, a B-box type 2, and a coiled-coil region. The protein localizes to both the nucleus and the cytoplasm, and may represent a participant in the initiation of glycogen synthesis. Multiple transcript variants have been found for this gene, and some of them encode the same isoform.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF90
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Trim7
Gene Accession No. Q9C029
Gene Alias AI790312; E3 ubiquitin-protein ligase TRIM7; Glycogenin-interacting protein; GNIP; RING finger protein 90; RNF90; TRIM7; tripartite motif containing 7; tripartite motif protein 7; tripartite motif protein TRIM7; tripartite motif-containing 7; tripartite motif-containing protein 7
Gene Symbols Trim7
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 67-95 amino acids from the N-terminal region of human TRIM7.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81786
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.