Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNPC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735086 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5086 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5086 Supplier Proteintech Group Inc Supplier No. 258201AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

U11/U12 snRNP 65K, also known as RBM40 (RNA-binding protein 40), RNPC3 (RNA-binding region-containing protein 3) or RNP, is a 517 amino acid nuclear protein that plays a role in pre-mRNA U12-dependent splicing by the minor spliceosome, ultimately leading to the removal of U12-type introns. Expressed at highest levels in kidney and pancreas, U11/U12 snRNP 65K is also weakly detected in placenta, heart, brain, lung, liver, thymus, colon, ovary, testis, prostate, spleen and small intestine. The gene encoding U11/U12 snRNP 65K maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome. Chromosome 1 houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson′s disease, Gaucher disease, schizophrenia and Usher syndrome.

Specifications

Antigen RNPC3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RNPC3
Gene Accession No. Q3UZ01, Q96LT9
Gene Alias KIAA183, KIAA1839, RBM40, RNA binding motif protein 40, RNA binding protein 40, RNP, RNPC3, U11/U12 65K, U11/U12 snRNP 65 kDa protein
Gene Symbols RNPC3
Host Species Rabbit
Immunogen RNPC3 Fusion Protein Ag22957
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55599, 67225
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.