Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ro 106-9920, Tocris Bioscience™

Catalog No. 177850
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-785-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-785-0 Supplier R&D Systems Supplier No. 1778/50
This item is not returnable. View return policy

Inhibitor of NF-κB activation

Inhibitor of NF-κB activation, possibly via selective inhibition of LPS- and TNF-α-induced IκBα ubiquitination (IC50 = 3 μM). Blocks subsequent production of TNF-α, IL-1β and IL-6. Inhibits mucin production in an in vitro model of COPD, and is anti-inflammatory following oral administration in vivo. Also weakly inhibits EGFR, 5-lipoxygenase and iNOS.

Chemical Identifiers

CAS 62645-28-7
Molecular Formula C10H7N5OS
Molecular Weight (g/mol) 245.26
InChI Key JFSXSNSCPNFCDM-UHFFFAOYSA-N
Synonym 6-benzenesulfinyl tetrazolo 1,5-b pyridazine, tetrazolo 1,5-b pyridazine, 6-phenylsulfinyl, 6-benzenesulfinyl-1,2,3,4 tetrazolo 1,5-b pyridazine, tocris-1778, tetrazolopyridazine-phenylsulfoxide, 6-phenylsulfinyl tetrazolo 1,5-b pyridazine
PubChem CID 3906779
IUPAC Name 6-(benzenesulfinyl)tetrazolo[1,5-b]pyridazine
SMILES C1=CC=C(C=C1)S(=O)C2=NN3C(=NN=N3)C=C2

Specifications

Quantity 50 mg
Formula Weight 245.26
Percent Purity >99%
Chemical Name or Material Ro 106-9920
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.