Learn More
Selleck Chemical LLC Ro 20-1724-E0346-25MG
SDP
Supplier:  Selleck Chemical LLC E034625MG
Encompass_Preferred
Ro 20-1724 is a potent inhibitor of PDE IV with Ki value of 3 1 M and a potent gastric secretagogues
Catalog No. 50-313-6941
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.