Learn More
TARGETMOL CHEMICALS INC RO 25-6981 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T12758
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ro 25-6981 is a potent and selective activity-dependent NMDA receptors blocker containing the NR2B subunit with IC50s of 0.009 and 52 uM for cloned receptor subunit combinations NR1C/NR2B and NR1C/NR2A respectively. Ro 25-6981 can be used in studies about Parkinson's disease. purity: 99%
Catalog No. 50-257-7415
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.