Learn More
Apexbio Technology LLC Ro 31-8220 Mesylate(Synonyms: Ro 31-8220 Methanesulfonate, Bisindolylmaleimide IX mesylate, Ro-318220 Mesylate), 10mM (in 1mL DMSO), CAS: 138489-18-6.
SDP
Supplier:  Apexbio Technology LLC B21925.1
Encompass_Preferred
Ro 31-8220 Mesylate (CAS 138489-18-6) is a selective small molecule inhibitor of protein kinase C (PKC) targeting multiple PKC isoforms ( I II and ) with IC50 values ranging from 5 to 27 nM PKC constitutes a family of calcium- and phospholipid-dependent serine/threonine kinases implicated in signaling pathways associated with growth factor responses and malignant transformation Ro 31-8220 modulates downstream targets inhibiting MKP-1 and c-Fos expression while inducing c-Jun and activating Jun N-terminal kinase (JNK) in a PKC-dependent manner It suppresses mitogen-induced IL-2 production and T-cell proliferation suggesting utility in immunological research and affects glucose metabolism via JNK and glycogen synthase activation relevant to metabolic studies
Catalog No. 50-000-60097
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.