Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RO-3306 95.0+%, TCI America™
SDP

Catalog No. R020150MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R020150MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R020150MG Supplier TCI America Supplier No. R020150MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 872573-93-8
Molecular Formula C18H13N3OS2
Molecular Weight (g/mol) 351.442
MDL Number MFCD17392573
InChI Key XOLMRFUGOINFDQ-YBEGLDIGSA-N
Synonym cdk1 inhibitor iv, ro-3306, 5-6-quinolinylmethylene-2-2-thienylmethyl amino-4 5h-thiazolone, 5z-5-quinolin-6-ylmethylidene-2-thiophen-2-ylmethyl amino-1,3-thiazol-4-one, z-5-quinolin-6-ylmethylene-2-thiophen-2-ylmethylamino thiazol-4 5h-one, ro hplc, z-5-quinolin-6-ylmethylene-2-thiophen-2-ylmethyl amino thiazol-4 5h-one, 2-thiophen-2-ylmethyl-amino-5-1-quinolin-6-yl-meth-z-ylidene-thiazol-4-one, 5z-5-quinolin-6-yl methylidene-2-thiophen-2-yl methyl amino-4,5-dihydro-1,3-thiazol-4-one
PubChem CID 11631681
IUPAC Name (5Z)-5-(quinolin-6-ylmethylidene)-2-(thiophen-2-ylmethylamino)-1,3-thiazol-4-one
SMILES C1=CC2=C(C=CC(=C2)C=C3C(=O)N=C(S3)NCC4=CC=CS4)N=C1

Specifications

Quantity 50 mg
Formula Weight 351.44
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material RO-3306
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.