accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 872573-93-8

IUPAC Name: 5-[(quinolin-6-yl)methylidene]-2-{[(thiophen-2-yl)methyl]amino}-4,5-dihydro-1,3-thiazol-4-one
Synonyms: 5-(quinolin-6-ylmethylidene)-2-[(thiophen-2-ylmethyl)amino]-1,3-thiazol-4-one
Molecular Weight (g/mol): 351.44
Molecular Formula: C18H13N3OS2
InChi Key: XOLMRFUGOINFDQ-UHFFFAOYSA-N
SMILES: O=C1N=C(NCC2=CC=CS2)SC1=CC1=CC2=CC=CN=C2C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results