Learn More
Medchemexpress LLC HY-19149 5mg , Ro24-7429 CAS:139339-45-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY191495MG
Encompass_Preferred
HY-19149 5mg Ro24-7429 CAS: 139339-45-0 Ro24-7429 is a potent and orally active HIV-1 transactivator protein Tat antagonist. Ro24-7429 is also a runt-related transcription factor 1 (RUNX1) inhibitor. Ro24-7429 has anti-HIV, antifibrotic and anti-inflammatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1904