Learn More
Invitrogen™ ROBO1 Monoclonal Antibody (10E2)

Description
ROBO1 Monoclonal Antibody for IHC (P), Western Blot
Specifications
Specifications
| Antigen | ROBO1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 10E2 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ROBO1 |
| Gene Accession No. | O89026, Q9Y6N7 |
| Gene Alias | AW494633; AW742721; deleted in U twenty twenty; DUTT1; FLJ21882; Gm310; HGNC:10249; H-Robo-1; MGC131599; MGC133277; ROBO1; roundabout 1; roundabout guidance receptor 1; roundabout homolog 1; roundabout homolog 1 (Drosophila); roundabout, axon guidance receptor, homolog 1; roundabout, axon guidance receptor, homolog 1 (Drosophila); Roundabout1 protein; SAX3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.