Learn More
ROBO1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
Specifications
Specifications
| Antigen | ROBO1 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | ROBO1 |
| Gene Accession No. | Q9Y6N7 |
| Gene Alias | ROBO1; DUTT1; roundabout, axon guidance receptor, homolog 1 (Drosophila) ; HGNC:10249; FLJ21882; SAX3; axon guidance receptor; roundabout 1; MGC131599; MGC133277 |
| Gene Symbols | ROBO1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.