Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ROBO1 Pre-design Chimera RNAi

Catalog No. 89035717 Shop All Abnova Corporation Products
Change view
Click to view available options
Description:
Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for 2 transcript variants. (NM_001145845.1, NM_133631.3)
Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for 2 transcript variants. (NM_002941.3,NM_133631.3)
Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for all of 3 transcript variants. (NM_001145845.1,NM_002941.3,NM_133631.3)
Quantity:
10 nmol
20 nmol
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Quantity
89-035-717 Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for 2 transcript variants. (NM_001145845.1, NM_133631.3) 20 nmol
89-035-712 Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for all of 3 transcript variants. (NM_001145845.1,NM_002941.3,NM_133631.3) 10 nmol
89-035-713 Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for all of 3 transcript variants. (NM_001145845.1,NM_002941.3,NM_133631.3) 20 nmol
89-035-714 Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for 2 transcript variants. (NM_002941.3,NM_133631.3) 10 nmol
89-035-715 Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for 2 transcript variants. (NM_002941.3,NM_133631.3) 20 nmol
89-035-716 Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for 2 transcript variants. (NM_001145845.1, NM_133631.3) 10 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 89-035-717 Supplier Abnova™ Supplier No. H00006091R05L

missing translation for 'validMainframeMsg'

Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for all of 3 transcript variants (NM_001145845.1,NM_002941.3,NM_133631.3)

  • Gene name: ROBO1
  • Gene Alias: DUTT1, FLJ21882, MGC131599, MGC133277, SAX3
  • Entrez GeneID: 6091
  • strong>Gene Description: Roundabout, axon guidance receptor, homolog 1

Specifications

Content And Storage Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity.
Description Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for 2 transcript variants. (NM_001145845.1, NM_133631.3)
Species Human
For Use With (Application) RNAi Knockdown
Includes DEPC water
Product Type ROBO1 Pre-design Chimera RNAi
Quantity 20 nmol
Target Human

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.