Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROBO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144712
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144712 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144712 Supplier Invitrogen™ Supplier No. PA5144712
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HLDELTRAYQ FDIAKQTWHI QSNNQPPQPP VPPLGYVSGA LISDLETDVA DDDADDEEEA LEIPRPLRAL DQTPGSSMDN LDSSVTGKAF TSSQRPRPTS PFSTDSNTSA ALSQSQRPRP TKKHKGGRMD QQPALPHRRE GM Antigen sequence identity: Mouse 92%; Rat 92%.

ROBO2 belongs to the ROBO family, part of the immunoglobulin superfamily proteins that are highly conserved from fly to human. ROBO2 is a receptor for SLIT2, molecules known to function in axon guidance and cell migration. Defects in this gene are the cause of vesicoureteral reflux type 2. Alternatively spliced transcript variants encoding different isoforms have been described for ROBO2.This gene belongs to the ROBO family, part of the immunoglobulin superfamily proteins that are highly conserved from fly to human. The encoded protein is a receptor for SLIT2, molecules known to function in axon guidance and cell migration. Defects in this gene are the cause of vesicoureteral reflux type 2. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROBO2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ROBO2
Gene Accession No. Q9HCK4
Gene Alias 2600013A04Rik; 9430089E08Rik; ast; astray; BB097918; D230004I22Rik; KIAA1568; mKIAA1568; ROBO2; roundabout 2; roundabout guidance receptor 2; roundabout homolog 2; roundabout homolog 2 (Drosophila); roundabout homolog 2-like; roundabout, axon guidance receptor, homolog 2; roundabout, axon guidance receptor, homolog 2 (Drosophila); roundabout2; SAX3; sb:eu937; Unknown (protein for IMAGE:8049638); unp1881; zmp:0000000549
Gene Symbols ROBO2
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 1168-1309. Recombinant protein control fragment (Product #RP-109728).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6092
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.